Tisk…

2-Fluoro-5-methylnicotinic acid 95+%

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-Fluoro-5-methylpyridine-3-carbxylic acid

PC535027-100MG PC535027-250MG
APOSPC535027-100MGEA 1370 CZK
APOSPC535027-100MG APOSPC535027-250MG
2-Fluoro-5-methylnicotinic acid 95+%
2-Fluoro-5-methylnicotinic acid
Vzorec: C₇H₆FNO₂
MW: 155,126 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11848712
CAS číslo: 1042986-00-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR