6-Chloro-3-(trifluoromethyl)pyridin-2-amine 95+%
Dodavatel: Apollo Scientific
|
Varování
|
PC535026-1G
PC535026-250MG
APOSPC535026-1GEA
8010
CZK
APOSPC535026-1G
APOSPC535026-250MG
6-Chloro-3-(trifluoromethyl)pyridin-2-amine 95+%
6-Chloro-3-(trifluoromethyl)pyridin-2-amine
Vzorec:
C₆H₄ClF₃N₂ MW: 196,558 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD19707620 CAS číslo: 79456-27-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...