3-Bromo-2-methyl-5-(trifluoromethyl)pyridine 95+%
Dodavatel: Apollo Scientific
|
Varování
|
PC535025-1G
PC535025-250MG
APOSPC535025-1GEA
21630
CZK
APOSPC535025-1G
APOSPC535025-250MG
3-Bromo-2-methyl-5-(trifluoromethyl)pyridine 95+%
3-Bromo-2-methyl-5-(trifluoromethyl)pyridine
Vzorec:
C₇H₅BrF₃N MW: 240,021 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD13193665 CAS číslo: 1211542-21-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...