Tisk…

6-Fluoro-2-methylnicotinaldehyde 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: 6-Fluoro-2-methylpyridine-3-carboxaldehyde

PC535023-1G PC535023-250MG
APOSPC535023-1GEA 6350 CZK
APOSPC535023-1G APOSPC535023-250MG
6-Fluoro-2-methylnicotinaldehyde 97%
6-Fluoro-2-methylnicotinaldehyde
Vzorec: C₇H₆FNO
MW: 139.127 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD05662399
CAS číslo: 884494-96-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR