4-Chloro-2-fluoro-3-methylbenzoic acid 97%
Dodavatel: Apollo Scientific
|
Varování
|
PC535017-100MG
PC535017-250MG
APOSPC535017-100MGEA
787
CZK
APOSPC535017-100MG
APOSPC535017-250MG
4-Chloro-2-fluoro-3-methylbenzoic acid 97%
4-Chloro-2-fluoro-3-methylbenzoic acid
Vzorec:
C₈H₆ClFO₂ Storage Temperature: Ambient |
MDL Number:
MFCD17169808 CAS číslo: 153556-55-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...