4-(Aminomethyl)-3,5-difluorobenzonitrile 97%
Dodavatel: Apollo Scientific
|
Varování
|
PC535013-100MG
PC535013-250MG
APOSPC535013-100MGEA
6580
CZK
APOSPC535013-100MG
APOSPC535013-250MG
4-(Aminomethyl)-3,5-difluorobenzonitrile 97%
4-(Aminomethyl)-3,5-difluorobenzonitrile
Vzorec:
C₈H₆F₂N₂ MW: 168,143 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD16876870 CAS číslo: 633336-81-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...