5-Chloro-2,3-difluorobenzaldehyde 97%
Dodavatel: Apollo Scientific
|
Varování
|
PC535012-1G
PC535012-250MG
APOSPC535012-1GEA
3640
CZK
APOSPC535012-1G
APOSPC535012-250MG
5-Chloro-2,3-difluorobenzaldehyde 97%
5-Chloro-2,3-difluorobenzaldehyde
Vzorec:
C₇H₃ClF₂O MW: 176,548 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD20924197 CAS číslo: 1783512-25-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...