Methyl 2-(2-fluoro-4-hydroxyphenyl)acetate 95+%
Dodavatel: Apollo Scientific
|
Varování
|
PC535005-1G
PC535005-250MG
APOSPC535005-1GEA
3540
CZK
APOSPC535005-1G
APOSPC535005-250MG
Methyl 2-(2-fluoro-4-hydroxyphenyl)acetate 95+%
Methyl 2-(2-fluoro-4-hydroxyphenyl)acetate
Vzorec:
C₉H₉FO₃ MW: 184,164 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18399286 CAS číslo: 91361-59-0 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...