trans-2-Fluorocyclopropanamine hydrochloride 95+%
Dodavatel: Apollo Scientific
|
Varování
|
PC535003-100MG
PC535003-250MG
APOSPC535003-100MGEA
2320
CZK
APOSPC535003-100MG
APOSPC535003-250MG
trans-2-Fluorocyclopropanamine hydrochloride 95+%
trans-2-Fluorocyclopropanamine hydrochloride
Vzorec:
C₃H₆FN·ClH MW: 111,546 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD28100743 CAS číslo: 114152-96-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...