7-Bromo-6-fluoro-2H-3,1-benzoxazine-2,4(1H)-dione
Dodavatel: Apollo Scientific
|
Varování
|
PC53473-1G
PC53473-250MG
APOSPC53473-1GEA
15940
CZK
APOSPC53473-1G
APOSPC53473-250MG
7-Bromo-6-fluoro-2H-3,1-benzoxazine-2,4(1H)-dione
7-Bromo-6-fluoro-2H-3,1-benzoxazine-2,4(1H)-dione
Vzorec:
C₈H₃BrFNO₃ MW: 260,017 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD30723301 CAS číslo: 1887002-15-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...