Tisk…

2,2,3,3-Tetrafluoro-6-nitro-1,4-benzodioxane

Dodavatel: Apollo Scientific
Varování

Synonyms: 2,2,3,3-Tetrafluoro-6-nitro-2,3-dihydro-1,4-benzodioxine

PC53470-1G PC53470-250MG
APOSPC53470-1GEA 17990 CZK
APOSPC53470-1G APOSPC53470-250MG
2,2,3,3-Tetrafluoro-6-nitro-1,4-benzodioxane
2,2,3,3-Tetrafluoro-6-nitro-1,4-benzodioxane
Vzorec: C₈H₃F₄NO₄
MW: 253,107 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30723299
CAS číslo: 89586-09-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR