3-Bromo-2-(trifluoromethyl)benzaldehyde 95%
Dodavatel: Apollo Scientific
|
Varování
|
PC53449-1G
PC53449-250MG
PC53449-5G
APOSPC53449-1GEA
6220
CZK
APOSPC53449-1G
APOSPC53449-250MG
APOSPC53449-5G
3-Bromo-2-(trifluoromethyl)benzaldehyde 95%
3-Bromo-2-(trifluoromethyl)benzaldehyde
Vzorec:
C₈H₄BrF₃O Storage Temperature: Ambient |
MDL Number:
MFCD22566297 CAS číslo: 1289057-68-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...