Tisk…

tert-Butyl (5-fluoro-1,3-thiazol-2-yl)carbamate

Dodavatel: Apollo Scientific

PC53419-100MG PC53419-1G PC53419-250MG
APOSPC53419-100MGEA 6020 CZK
APOSPC53419-100MG APOSPC53419-1G APOSPC53419-250MG
tert-Butyl (5-fluoro-1,3-thiazol-2-yl)carbamate
tert-Butyl (5-fluoro-1,3-thiazol-2-yl)carbamate
Vzorec: C₈H₁₁FN₂O₂S
MW: 218,249 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11656630
CAS číslo: 731018-54-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR