Tisk…

Methyl 2-bromo-4-(trifluoromethyl)benzoate

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-Bromo-4-(trifluoromethyl)benzoic acid methyl ester

PC53418-1G PC53418-25G PC53418-5G PC53418-10G
APOSPC53418-1GEA 622 CZK
APOSPC53418-1G APOSPC53418-25G APOSPC53418-5G APOSPC53418-10G
Methyl 2-bromo-4-(trifluoromethyl)benzoate
Methyl 2-bromo-4-(trifluoromethyl)benzoate
Vzorec: C₉H₆BrF₃O₂
MW: 283,042 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD14697995
CAS číslo: 1214334-90-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR