2,4-Difluoro-5-iodobenzyl alcohol 95%
Dodavatel: Apollo Scientific
|
Varování
|
PC53417-1G
PC53417-250MG
PC53417-5G
APOSPC53417-1GEA
4630
CZK
APOSPC53417-1G
APOSPC53417-250MG
APOSPC53417-5G
2,4-Difluoro-5-iodobenzyl alcohol 95%
2,4-Difluoro-5-iodobenzyl alcohol
Vzorec:
C₇H₅F₂IO Storage Temperature: Ambient |
MDL Number:
MFCD28348701 CAS číslo: 1097626-21-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...