Tisk…

4-Bromo-5-fluoro-2-iodobenzonitrile 95%

Dodavatel: Apollo Scientific
Varování

PC53385-1G PC53385-250MG PC53385-5G
APOSPC53385-1GEA 12010 CZK
APOSPC53385-1G APOSPC53385-250MG APOSPC53385-5G
4-Bromo-5-fluoro-2-iodobenzonitrile 95%
4-Bromo-5-fluoro-2-iodobenzonitrile
Vzorec: C₇H₂BrFIN
MW: 325,904 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30297368
CAS číslo: 2149601-45-4
UN: 3439
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR