Tisk…

4-(1,1-Difluoroethyl)benzyl alcohol

Dodavatel: Apollo Scientific
Varování

PC53355-1G PC53355-250MG
APOSPC53355-1GEA 24600 CZK
APOSPC53355-1G APOSPC53355-250MG
4-(1,1-Difluoroethyl)benzyl alcohol
4-(1,1-Difluoroethyl)benzyl alcohol
Vzorec: C₉H₁₀F₂O
Storage Temperature: Refrigerator
MDL Number: MFCD27925457
CAS číslo: 55805-25-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR