Tisk…

Ethyl 4-bromo-3-fluorobenzoate 99%

Dodavatel: Apollo Scientific
Varování

PC53348-1G PC53348-5G PC53348-25G
APOSPC53348-1GEA 622 CZK
APOSPC53348-1G APOSPC53348-5G APOSPC53348-25G
Ethyl 4-bromo-3-fluorobenzoate 99%
Ethyl 4-bromo-3-fluorobenzoate
Vzorec: C₉H₈BrFO₂
MW: 247,061 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09751983
CAS číslo: 1130165-74-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR