Tisk…

2-Bromo-5-chloro-4-fluorotoluene

Dodavatel: Apollo Scientific
Varování

Synonyms: 1-Bromo-4-chloro-5-fluoro-2-methylbenzene

PC53331-1G PC53331-250MG
APOSPC53331-1GEA 4140 CZK
APOSPC53331-1G APOSPC53331-250MG
2-Bromo-5-chloro-4-fluorotoluene
2-Bromo-5-chloro-4-fluorotoluene
Vzorec: C₇H₅BrClF
Storage Temperature: Ambient
MDL Number: MFCD20441895
CAS číslo: 1067882-53-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR