Tisk…

4-Bromo-2-fluoro-6-methylbenzoic acid 95%

Dodavatel: Apollo Scientific
Varování

PC53324-1G PC53324-5G PC53324-10G
APOSPC53324-1GEA 992 CZK
APOSPC53324-1G APOSPC53324-5G APOSPC53324-10G
4-Bromo-2-fluoro-6-methylbenzoic acid 95%
4-Bromo-2-fluoro-6-methylbenzoic acid
Vzorec: C₈H₆BrFO₂
MW: 233,034 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18072908
CAS číslo: 1242157-23-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR