Tisk…

Methyl 4-bromo-5-fluoro-2-hydroxybenzoate 95%

Dodavatel: Apollo Scientific
Varování

PC53322-1G PC53322-5G PC53322-10G
APOSPC53322-1GEA 953 CZK
APOSPC53322-1G APOSPC53322-5G APOSPC53322-10G
Methyl 4-bromo-5-fluoro-2-hydroxybenzoate 95%
Methyl 4-bromo-5-fluoro-2-hydroxybenzoate
Vzorec: C₈H₆BrFO₃
Storage Temperature: Ambient
MDL Number: MFCD23701539
CAS číslo: 1193162-25-2

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR