Tisk…

4-Bromo-5-chloro-2-fluorobenzenesulphonyl chloride 97%

Dodavatel: Apollo Scientific
Nebezpečí

PC53319-1G PC53319-5G PC53319-10G PC53319-25G
APOSPC53319-1GEA 787 CZK
APOSPC53319-1G APOSPC53319-5G APOSPC53319-10G APOSPC53319-25G
4-Bromo-5-chloro-2-fluorobenzenesulphonyl chloride 97%
4-Bromo-5-chloro-2-fluorobenzenesulphonyl chloride
Vzorec: C₆H₂BrCl₂FO₂S
Storage Temperature: Ambient
MDL Number: MFCD12026216
CAS číslo: 1208075-41-5
UN: 3261
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR