Tisk…

6-Bromo-2-fluoro-3-(trifluoromethoxy)phenol

Dodavatel: Apollo Scientific
Varování

PC53314-1G PC53314-250MG
APOSPC53314-1GEA 6620 CZK
APOSPC53314-1G APOSPC53314-250MG
6-Bromo-2-fluoro-3-(trifluoromethoxy)phenol
6-Bromo-2-fluoro-3-(trifluoromethoxy)phenol
Vzorec: C₇H₃BrF₄O₂
MW: 274.995 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28740790
CAS číslo: 1805580-01-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR