1-(3-Bromo-4-fluorophenyl)-2,2,2-trifluoroethanol
Dodavatel: Apollo Scientific
|
Varování
|
PC53306-1G
PC53306-250MG
APOSPC53306-1GEA
10150
CZK
APOSPC53306-1G
APOSPC53306-250MG
1-(3-Bromo-4-fluorophenyl)-2,2,2-trifluoroethanol
1-(3-Bromo-4-fluorophenyl)-2,2,2-trifluoroethanol
Vzorec:
C₈H₅BrF₄O MW: 273,022 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD16067892 CAS číslo: 150698-79-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...