Difluoromethanesulphonyl chloride
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC53298-1G
PC53298-5G
PC53298-250MG
APOSPC53298-1GEA
5630
CZK
APOSPC53298-1G
APOSPC53298-5G
APOSPC53298-250MG
Difluoromethanesulphonyl chloride
Difluoromethanesulphonyl chloride
Vzorec:
CHClF₂O₂S Bod varu: 95…99 °C Hustota: 1,696 g/cm³ (25 °C) Bod vzplanutí: 5 °C Storage Temperature: Ambient |
MDL Number:
MFCD09971548 CAS číslo: 1512-30-7 EINECS: 216-150-7 UN: 2924 ADR: 3+8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...