Tisk…

2-(Bromomethyl)-5-fluoropyridine hydrobromide 95%

Dodavatel: Apollo Scientific
Varování

PC53288-1G PC53288-250MG
APOSPC53288-1GEA 45630 CZK
APOSPC53288-1G APOSPC53288-250MG
2-(Bromomethyl)-5-fluoropyridine hydrobromide 95%
2-(Bromomethyl)-5-fluoropyridine hydrobromide
Vzorec: C₆H₅BrFN·BrH
MW: 270,925 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12546970
CAS číslo: 1186194-88-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR