2-Fluoro-5-nitrobenzyl bromide 98%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC53278-1G
PC53278-25G
PC53278-5G
APOSPC53278-1GEA
2690
CZK
APOSPC53278-1G
APOSPC53278-25G
APOSPC53278-5G
2-Fluoro-5-nitrobenzyl bromide 98%
2-Fluoro-5-nitrobenzyl bromide
Vzorec:
C₇H₅BrFNO₂ Bod tání: 74…76 °C Storage Temperature: Ambient |
MDL Number:
MFCD07787098 CAS číslo: 454-15-9 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...