1-(1-Bromo-2,2,2-trifluoroethyl)-4-nitrobenzene
Dodavatel: Apollo Scientific
|
Varování
|
PC53275-1G
PC53275-250MG
APOSPC53275-1GEA
14880
CZK
APOSPC53275-1G
APOSPC53275-250MG
1-(1-Bromo-2,2,2-trifluoroethyl)-4-nitrobenzene
1-(1-Bromo-2,2,2-trifluoroethyl)-4-nitrobenzene
Vzorec:
C₈H₅BrF₃NO₂ MW: 284,03 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD29912987 CAS číslo: 2091158-96-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...