Tisk…

5-Fluoro-2-iodoanisole

Dodavatel: Apollo Scientific
Varování

Synonyms: 4-Fluoro-1-iodo-2-methoxybenzene

PC53271-1G PC53271-5G
APOSPC53271-1GEA 3440 CZK
APOSPC53271-1G APOSPC53271-5G
5-Fluoro-2-iodoanisole
5-Fluoro-2-iodoanisole
Vzorec: C₇H₆FIO
MW: 252,025 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18396916
CAS číslo: 450-90-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR