4-(Difluoromethoxy)-2-fluorobenzaldehyde
Dodavatel: Apollo Scientific
|
Varování
|
PC53249-1G
PC53249-5G
APOSPC53249-1GEA
1580
CZK
APOSPC53249-1G
APOSPC53249-5G
4-(Difluoromethoxy)-2-fluorobenzaldehyde
4-(Difluoromethoxy)-2-fluorobenzaldehyde
Vzorec:
C₈H₅F₃O₂ MW: 190.119 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11847162 CAS číslo: 1214358-05-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...