3-Fluoro-4-sulphamoylbenzoic acid
Dodavatel: Apollo Scientific
|
Varování
|
PC53203-1G
PC53203-250MG
APOSPC53203-1GEA
6120
CZK
APOSPC53203-1G
APOSPC53203-250MG
3-Fluoro-4-sulphamoylbenzoic acid
3-Fluoro-4-sulphamoylbenzoic acid
Vzorec:
C₇H₆FNO₄S MW: 219,19 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD21602650 CAS číslo: 244606-37-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...