4-Fluorobenzenesulphinic acid sodium salt
Dodavatel: Apollo Scientific
|
Varování
|
PC53199-25G
PC53199-5G
APOSPC53199-25GEA
695
CZK
APOSPC53199-25G
APOSPC53199-5G
4-Fluorobenzenesulphinic acid sodium salt
4-Fluorobenzenesulphinic acid sodium salt
Vzorec:
C₆H₄FO₂S·Na Storage Temperature: Ambient |
MDL Number:
MFCD03093775 CAS číslo: 824-80-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...