Tisk…

3-Bromo-2-(trifluoromethyl)benzoic acid

Dodavatel: Apollo Scientific
Varování

PC53198-1G PC53198-250MG PC53198-5G
APOSPC53198-1GEA 4300 CZK
APOSPC53198-1G APOSPC53198-250MG APOSPC53198-5G
3-Bromo-2-(trifluoromethyl)benzoic acid
3-Bromo-2-(trifluoromethyl)benzoic acid
Vzorec: C₈H₄BrF₃O₂
MW: 269.015 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16606443
CAS číslo: 1227605-02-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR