2-Amino-4,6-difluorobenzonitrile 95%
Dodavatel: Apollo Scientific
|
Varování
|
PC53175-1G
PC53175-5G
APOSPC53175-1GEA
2240
CZK
APOSPC53175-1G
APOSPC53175-5G
2-Amino-4,6-difluorobenzonitrile 95%
2-Amino-4,6-difluorobenzonitrile
Vzorec:
C₇H₄F₂N₂ MW: 154,117 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD13178245 CAS číslo: 190011-84-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...