Tisk…

2-Fluoropropanedioic acid 95%

Dodavatel: Apollo Scientific
Nebezpečí

PC53171-1G PC53171-250MG
APOSPC53171-1GEA 9330 CZK
APOSPC53171-1G APOSPC53171-250MG
2-Fluoropropanedioic acid 95%
2-Fluoropropanedioic acid
Vzorec: C₃H₃FO₄
Storage Temperature: Ambient
MDL Number: MFCD19228375
CAS číslo: 473-87-0
UN: 3261
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR