2-Chloro-6-fluoro-4-nitroaniline
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC53167-1G
PC53167-5G
APOSPC53167-1GEA
4400
CZK
APOSPC53167-1G
APOSPC53167-5G
2-Chloro-6-fluoro-4-nitroaniline
2-Chloro-6-fluoro-4-nitroaniline
Vzorec:
C₆H₄ClFN₂O₂ MW: 190.56 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD23135899 CAS číslo: 350-20-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...