3,5-Difluoro-4-hydroxyacetophenone 99%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
1-(3,5-Difluoro-4-hydroxyphenyl)ethanone
PC53154-250MG
PC53154-100MG
APOSPC53154-250MGEA
19010
CZK
APOSPC53154-250MG
APOSPC53154-100MG
3,5-Difluoro-4-hydroxyacetophenone 99%
3,5-Difluoro-4-hydroxyacetophenone
Vzorec:
C₈H₆F₂O₂ MW: 172,129 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD09787482 CAS číslo: 133186-55-7 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...