Tisk…

2-(2-Fluoro-6-nitrophenyl)acetic acid

Dodavatel: Apollo Scientific
Varování

PC53138-1G PC53138-250MG
APOSPC53138-1GEA 5390 CZK
APOSPC53138-1G APOSPC53138-250MG
2-(2-Fluoro-6-nitrophenyl)acetic acid
2-(2-Fluoro-6-nitrophenyl)acetic acid
Vzorec: C₈H₆FNO₄
MW: 199.136 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13185636
CAS číslo: 136916-19-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR