6-Bromo-4-chloro-8-fluoroquinoline 97%
Dodavatel: Apollo Scientific
|
Varování
|
PC53129-1G
PC53129-250MG
PC53129-100MG
APOSPC53129-1GEA
8010
CZK
APOSPC53129-1G
APOSPC53129-250MG
APOSPC53129-100MG
6-Bromo-4-chloro-8-fluoroquinoline 97%
6-Bromo-4-chloro-8-fluoroquinoline
Vzorec:
C₉H₄BrClFN MW: 260,49 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09861346 CAS číslo: 1019016-66-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...