5-Fluoro-3-cyclopropylphenol
Dodavatel: Apollo Scientific
|
Varování
|
PC53081-1G
PC53081-250MG
APOSPC53081-1GEA
2070
CZK
APOSPC53081-1G
APOSPC53081-250MG
5-Fluoro-3-cyclopropylphenol
5-Fluoro-3-cyclopropylphenol
Vzorec:
C₉H₉FO MW: 152,166 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD25994491 CAS číslo: 1402666-87-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...