Tisk…

2-(Difluoromethoxy)-5-fluorobromobenzene 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-Bromo-1-(difluoromethoxy)-4-fluorobenzene

PC53074-1G PC53074-250MG PC53074-5G
APOSPC53074-1GEA 18020 CZK
APOSPC53074-1G APOSPC53074-250MG APOSPC53074-5G
2-(Difluoromethoxy)-5-fluorobromobenzene 97%
2-(Difluoromethoxy)-5-fluorobromobenzene
Vzorec: C₇H₄BrF₃O
Storage Temperature: Ambient
MDL Number: MFCD09935620
CAS číslo: 936249-94-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR