2,6-Difluoro-N-methoxy-N-methyl-4-(trifluoromethyl)benzamide
Dodavatel: Apollo Scientific
|
Varování
|
PC53066-1G
PC53066-250MG
APOSPC53066-1GEA
32730
CZK
APOSPC53066-1G
APOSPC53066-250MG
2,6-Difluoro-N-methoxy-N-methyl-4-(trifluoromethyl)benzamide
2,6-Difluoro-N-methoxy-N-methyl-4-(trifluoromethyl)benzamide
Vzorec:
C₁₀H₈F₅NO₂ MW: 269,168 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD28963752 CAS číslo: 1980040-18-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...