Tisk…

5-Bromo-2-fluoro-4-methylphenyl trifluoromethanesulphonate

Dodavatel: Apollo Scientific
Nebezpečí

PC53047-1G PC53047-5G
APOSPC53047-1GEA 5360 CZK
APOSPC53047-1G APOSPC53047-5G
5-Bromo-2-fluoro-4-methylphenyl trifluoromethanesulphonate
5-Bromo-2-fluoro-4-methylphenyl trifluoromethanesulphonate
Vzorec: C₈H₅BrF₄O₃S
MW: 337,086 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28963743
CAS číslo: 1936290-01-5
UN: 3265
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR