Tisk…

5-Bromo-6-fluoroisatoic anhydride

Dodavatel: Apollo Scientific
Varování

PC53036-1G PC53036-5G
APOSPC53036-1GEA 12730 CZK
APOSPC53036-1G APOSPC53036-5G
5-Bromo-6-fluoroisatoic anhydride
5-Bromo-6-fluoroisatoic anhydride
Vzorec: C₈H₃BrFNO₃
Storage Temperature: Ambient
MDL Number: MFCD28897196
CAS číslo: 1935441-96-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR