Tisk…

5-Fluoro-2-(2,2,2-trifluoroethoxy)nitrobenzene

Dodavatel: Apollo Scientific
Varování

Synonyms: 4-Fluoro-2-nitro-1-(2,2,2-trifluoroethoxy)benzene, 4-Fluoro-2-nitrophenyl 2,2,2-trifluoroethyl ether, 2-Nitro-β,β,β,4-tetrafluorophenetole

PC53031-1G PC53031-5G
APOSPC53031-1GEA 11580 CZK
APOSPC53031-1G APOSPC53031-5G
5-Fluoro-2-(2,2,2-trifluoroethoxy)nitrobenzene
5-Fluoro-2-(2,2,2-trifluoroethoxy)nitrobenzene
Vzorec: C₈H₅F₄NO₃
Storage Temperature: Ambient
MDL Number: MFCD28155023
CAS číslo: 334929-98-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR