Tisk…

2-Amino-4-bromo-6-fluorobenzonitrile

Dodavatel: Apollo Scientific
Varování

PC53029-1G PC53029-250MG PC53029-5G
APOSPC53029-1GEA 662 CZK
APOSPC53029-1G APOSPC53029-250MG APOSPC53029-5G
2-Amino-4-bromo-6-fluorobenzonitrile
2-Amino-4-bromo-6-fluorobenzonitrile
Vzorec: C₇H₄BrFN₂
Storage Temperature: Ambient
MDL Number: MFCD28155022
CAS číslo: 1279865-14-3
UN: 3439
ADR: 6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR