3-Bromo-2-chloro-6-fluorophenacyl bromide
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC53028-1G
PC53028-5G
APOSPC53028-1GEA
13660
CZK
APOSPC53028-1G
APOSPC53028-5G
3-Bromo-2-chloro-6-fluorophenacyl bromide
3-Bromo-2-chloro-6-fluorophenacyl bromide
Vzorec:
C₈H₄Br₂ClFO Storage Temperature: Ambient |
MDL Number:
MFCD28155021 CAS číslo: 1805575-98-7 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...