3-Bromo-2-chloro-6-fluoroaniline, N-BOC protected
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
3-Bromo-N-(tert-butoxycarbonyl)-2-chloro-6-fluoroaniline, tert-Butyl (3-bromo-2-chloro-6-fluorophenyl)carbamate
PC53026-1G
PC53026-5G
APOSPC53026-1GEA
10250
CZK
APOSPC53026-1G
APOSPC53026-5G
3-Bromo-2-chloro-6-fluoroaniline, N-BOC protected
3-Bromo-2-chloro-6-fluoroaniline, N-BOC protected
Vzorec:
C₁₁H₁₂BrClFNO₂ Storage Temperature: Ambient |
MDL Number:
MFCD28140552 CAS číslo: 1820673-55-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...