Tisk…

4-Amino-3-fluorobenzyl bromide, N-BOC protected

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: tert-Butyl [4-(bromomethyl)-2-fluorophenyl]carbamate

PC53017-1G PC53017-250MG PC53017-5G
APOSPC53017-1GEA 14550 CZK
APOSPC53017-1G APOSPC53017-250MG APOSPC53017-5G
4-Amino-3-fluorobenzyl bromide, N-BOC protected
4-Amino-3-fluorobenzyl bromide, N-BOC protected
Vzorec: C₁₂H₁₅BrFNO₂
Storage Temperature: Ambient
MDL Number: MFCD11976469
CAS číslo: 323578-36-5
UN: 3261
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR