2'-(Trifluoromethyl)-biphenyl-4-methanamine hydrochloride 97%
Dodavatel: Apollo Scientific
PC530057-1G
PC530057-500MG
APOSPC530057-1GEA
18620
CZK
APOSPC530057-1G
APOSPC530057-500MG
2'-(Trifluoromethyl)-biphenyl-4-methanamine hydrochloride 97%
2'-(Trifluoromethyl)-biphenyl-4-methanamine hydrochloride
Vzorec:
C₁₄H₁₂F₃N·ClH MW: 287,708 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD31536877 CAS číslo: 472964-20-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...